Ismatec Standard Cartridge Pump Head for 2-Stop Tubing, 12-Channel, 8-Roller
Representative image only.
Ismatec Item # WZ-78002-36
Ideal pump-head for multichannel applications at higher flow rates
  • Snap your pump head onto your drives quickly with turn-and-lock mounting system
  • Select from sixteen interchangeable pump heads that offer flow rates from 0.01 to 3660 mL/min
  • RS-232 interface connects to your PC
  • Features 12 channels
PRODUCT OPTIONS
£1,585.18GBP / Each
£1,902.22GBP (incl VAT)
£118.10GBP / Each
£141.72GBP (incl VAT)
for an additional
£118.10GBP / Each
£141.72GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • SeriesIsmatec
  • Max Flow Rate (mL/min)180
  • Min Flow Rate (mL/min)0.01
  • Max Pressure (PSI)22
  • Number of Channels12
  • Rollers8
  • DescriptionStandard cartridge pump head, 12 Ch, 8 rollers, 2-stop
Key Features
  • Snap your pump head onto your drives quickly with turn-and-lock mounting system
  • Select from sixteen interchangeable pump heads that offer flow rates from 0.01 to 3660 mL/min
  • RS-232 interface connects to your PC
  • Features 12 channels
More About this Item
Select from four-, eight-, or twelve-channel pump heads. They deliver a wide range of flow rates—as low as 0.01 mL/min to as high as 180 mL/min! Click ‘n’ go cartridges allow quick and easy tubing changes.
The first of a series of two-stop tubing options is below under Accessories. See all of our two-stop tubing formulations or call our Applications Specialists for personal assistance.