This item is now discontinued. Please browse similar items

Interscience Laboratories 122-225 Bagpage Round Bottom Paddle Blender Bags, 400 mL With Full Filter, 280 microns

Representative image only.
Interscience Laboratories Mfr # 122-225 – Item # WZ-36850-29
  • Round bottom
  • Multilayer®, reinforced multicoated complex

SPECS & DESCRIPTION

Specifications & Description

  • Bag TypePaddle Blender Bags
  • Bag Capacity (mL)400
  • Bag Capacity (L)0.4
  • DescriptionBagpage Round Bottom Paddle Blender Bags, 400 mL With Full Filter, 280 microns
Conformite EuropeenneISO9001:2000 Certified Supplier

Key Features

  • Round bottom
  • Multilayer®, reinforced multicoated complex

More About this Item

Choose from a variety of bags to use with your paddle blender. Blending bags are available in various sizes and come with and without filters.

Recommended for solid sample preparation of thick, pasty products and solid sample preparation requiring filtration
Porosity of the filter: 280 microns

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat