This item is now discontinued. Please browse similar items
International Products Corp Z-0720 Zymit Pro Enzyme Cleaner, 21 KG (19 L)
Representative image only.
International Products Corp Mfr # Z-0720 – Item # WZ-18100-18
Biodegradable enzyme and surfactant cleaner that breaks down and removes proteinaceous soils.
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Dilution1:100 to 2:100, pH 7.5
  • ApplicationsBlood, Emulsifiers, Fingerprints, Fragrances, Gels, Proteins
  • Description Z-0720 ZYMIT PRO ENZYME CLEANER 21.0 KG NET WT CONTAINER
More About this Item
It is an effective filter membrane cleaner, especially when used with Micro-90 or Micro A07 in consecutive cleaning steps.