This item is now discontinued. View the direct replacement: WZ-18100-23 or browse similar items
International Products Corp M-1050-12 Micro Green Clean Biodegradable Cleaner, 12 x 1 L
Representative image only.
International Products Corp Mfr # M-1050-12 – Item # WZ-18100-08
A free-rinsing, industrial strength, biodegradable hard surface cleaner designed for use in a wide range of applications.
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)0.25
  • Volume (Liters)1
  • Dilution1:100 to 2:100, pH 9.8
  • ApplicationsGels, Emulsifiers, Oil, Titanium Doixides, Dyes, Silicaons, Fragrances, Petrolatum, Paraffins
  • Description M-1050-12 MICRO GREEN CLEAN BIODEGRADABLE CLEANER 1 LITER CONTAINER (12 PER CASE)
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Excellent for cleaning metals, ceramics, medical instruments, food-processing equipment, filter membranes, and other surfaces. Typical pH: 9.5.