This item is now discontinued. Please browse similar items
International Products Corp M-0820 Micro A07 Citric Acid Cleaner 20L bottle
Representative image only.
International Products Corp Mfr # M-0820 – Item # WZ-18104-30
Environmentally friendly – Phosphate Free
  • Free Rinsing
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)5
  • Volume (Liters)20
  • Dilution1:100 to 2:100, 2.5 pH
  • DescriptionMicro A07 Citric Acid Cleaner 20L bottle
Key Features
  • Free Rinsing
More About this Item
Micro A07 is a free-rinsing cleaner designed for use in a wide range of applications including ultrasonic baths, immersion tanks, mild-agitation systems, CIP systems and more. It is ideal for cleaning metals, stainless steel, ceramics, glass, plastics, and filter membranes. Micro A07 solution removes a variety of soils as well including scale, oxides, milkstone, mineral deposits, and inorganic materials. It is free of CFCs, ODCs, phosphates, silicates, borates, halogenated compounds, and phenols.