This item is now discontinued. Please browse similar items
International Enviroguard Sterile Cleanroom Coveralls, large, 25/cs
Representative image only.
International Enviroguard Mfr # CE11013CIS-L – Item # WZ-33684-32
Bound seams and tunnelized elastic provide low linting and strong tear resistance
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • SizeLarge
  • ColorWhite
  • ComplianceISO Class 4 (formerly Class 10)
  • DescriptionSterile Cleanroom Coveralls, large, 25/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Coveralls feature a sterility assurance of 10-6, low linting, and a more breathable liquid barrier. Special fold design protects against potential contamination during gowning. Feature bound seams and elastic wrists and ankles. Manufactured in an ISO certified facility, clean processed, and individually packaged under cleanroom conditions. Recyclable.
Compliance: ISO Class 4