This item is now discontinued. Please browse similar items
International Enviroguard PyroGuard FR Flame-Retardant Coveralls, Elastic Wrist, Hood/Boots, XXL; 25/cs
Representative image only.
International Enviroguard Mfr # 9019-2XL – Item # WZ-86234-68
Disposable coveralls that protect primary flame-resistant apparel
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Flame retardant, water repellent
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyester
  • SizeXX-large
  • ColorBlue
  • ComplianceNFPA 701
  • DescriptionPyroGuard FR Flame-Retardant Coveralls, Elastic Wrist, Hood/Boots, XXL; 25/cs
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Flame retardant, water repellent
More About this Item
Coveralls are made with a tough, breathable, and lightweight material constructed of a hydro-entangled wood pulp and polyester. The surface is treated with a phosphate-based flame retardant chemical. These coveralls are intended to be worn over primary flame-resistant apparel, protecting it from dirt, grease, and non-toxic liquids. Passes NFPA 701 for vertical flammability.