This item is now discontinued. Please browse similar items
International Enviroguard Cleanroom Coveralls, elastic wrist and ankle, large
Representative image only.
International Enviroguard Mfr # CE8013-CI-L – Item # WZ-33683-12
Provides superior worker comfort that is also strong and durable
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene(PP)
  • SizeLarge
  • ColorWhite
  • ComplianceISO Class 5 (formerly Class 100)
  • DescriptionCleanroom Coveralls, elastic wrist and ankle, large
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Constructed of a soft, non-woven polypropylene combined with a microporous film, coveralls provide a superior liquid barrier, particulate barrier, and strong tear resistance. The microporous film provides enhanced breathability while the non-woven fabric presents a soft interior for increased worker comfort. Bound seams and tunnelized elastic ensure extremely low linting. Clean processed and individually packaged.
Compliance: ISO Class 5