This item is now discontinued. View the direct replacement: WZ-17104-00 or browse similar items
InnoCal NIST-Traceable Calibration; Pipettors, with 4 Measurements at 3 Volumes
Representative image only.
InnoCal Item # WZ-17104-01
Ensure your instrument remains precise
  • Receive your product calibrated and ready to use
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • NIST-Traceable calibration for pipettors with 4 measurements
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeLiquid Testing
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points3 points, 4 aliquots each
  • Calibration Test Points20, 50 and 100% of full scale
  • Calibration Test Rangeup to 4000 mL
  • DescriptionNIST-Traceable Calibration with 4 measurements at 3 volumes
NIST Traceable Calibration
Key Features
  • Receive your product calibrated and ready to use
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • NIST-Traceable calibration for pipettors with 4 measurements
More About this Item
InnoCal is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA) under certificate number 1746.01. This accreditation is your assurance that the calibration is performed in a qualified, controlled laboratory.
Calibrations on pipettes, dispensers and graduated cylinders are performed per the procedures in ISO8655 using a precision pipette accuracy tester. Before testing, the valves and seals on pipettes and dispensers are tested for integrity using a leak tester. Variable pipettors and dispensers are measured at 3 points at 10, 50 and 100% of the maximum value. Fixed pipettors and dispensers are measured at the nominal value. Evaluation of the unit under test is done for dispensed volume and the coefficient of variability (CV) value.