This item is now discontinued. Please browse similar items
InnoCal NIST-Traceable Calibration; Temperature Transmitter SKU 1710308
Representative image only.
InnoCal Item # WZ-17103-08
NIST-traceable calibrations for transmitters, temperature baths, and scanning thermometers
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Not an accredited certificate
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeTemperature
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points4 points
  • Calibration Test PointsITS-90 points across the range unless specfied
  • Calibration Test Range-196 to 1300 C
  • DescriptionNIST-Traceable Calibration; Temperature Transmitter
Key Features
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Not an accredited certificate
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
InnoCal's Metrology Laboratory is accredited to ISO/IEC Guide 25 by the American Association for Laboratory Accreditation (A2LA); certificate number 1746.01. This accreditation is your assurance that one of the foremost outside auditing agencies approves of our internal laboratory policies and procedures.