This item is now discontinued. View the direct replacement: WZ-17101-53 or browse similar items
InnoCal NIST-Traceable Calibration; Sound-Level Calibrator
Representative image only.
InnoCal Item # WZ-17101-54
NIST-traceable sound calibrations--service available for sound-level meters (types I & II), noise dosimeters, sound filters, and sound-level calibrators
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeEnvironmental Monitoring
  • Instrument Only CalibrationYes
  • Number Of Calibration Test Points2 Points per Range of the instrument
  • DescriptionNIST-Traceable Calibration; Sound-Level Calibrator
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005