This item is now discontinued. View the direct replacement: WZ-17101-86 or browse similar items
InnoCal NIST-Traceable Calibration; Precision Torque Tester, 1,000 lb-ft Maximum
Representative image only.
InnoCal Item # WZ-17101-88
NIST-traceable calibration for torque wrenches, screwdrivers, testers, and more!
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeForce and Torque
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points5 Points Clockwise, 5 Points Counterclockwise
  • Calibration Test PointsThroughout the range
  • Calibration Test Range0 to 2000 ft-lbf
  • DescriptionNIST-Traceable Calibration; Precision Torque Tester, 1,000 lb-ft maximum
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005