InnoCal NIST-Traceable Calibration; Multiparameter Timer
Representative image only.
InnoCal Item # WZ-17060-01
Quick and accurate timer calibrations accredited to ISO/IEC 17025 standards
  • NIST-traceable calibration allows you to confirm that your equipment meets the manufacturer’s published specification EPA, USDA, ISO and GLP/GMPs standards
  • Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
PRODUCT OPTIONS
£165.05GBP / Each
£198.06GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeMechanical
  • System Calibration (Instrument and Probe)Yes
  • DescriptionNIST-Traceable for Multiparameter Timers
Key Features
  • NIST-traceable calibration allows you to confirm that your equipment meets the manufacturer’s published specification EPA, USDA, ISO and GLP/GMPs standards
  • Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
More About this Item
Calibrations on timers and stopwatches are performed using a precision timometer. Measurements can be taken from 60 seconds to 720 hours.
Calibrations on timers and stopwatches are performed using a precision timometer. Measurements can be taken from 60 seconds to 720 hours. If present, humidity functions are tested using a chilled mirror system and barometric functions are tested using high-precision quartz pressure reference transducers.