This item is now discontinued. Please browse similar items
InnoCal MSA Mini Single-Gas Calibration
Representative image only.
InnoCal Item # WZ-17102-13
Protect personnel from gas hazards due to inaccurate readings – calibrate your gas detector today!
  • Factory authorized service center for MSA and Sperian gas detectors
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeAir and Gas Measurement
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points1 per gas range
  • Calibration Test PointsPer manufacturer's recommendation
  • DescriptionMSA Mini Single-Gas Calibration
Key Features
  • Factory authorized service center for MSA and Sperian gas detectors
More About this Item
InnoCal's specially trained technicians test your gas detector with a known concentration of gas. Calibration drift can occur due to many factors: normal degradation of sensors over time, exposure to extreme temperature and humidity conditions, exposure to high levels of the target gases, and harsh storage and handling conditions such as being dropped or submerged in liquid. Ensure worker safety by having your gas detector calibrated by our factory-trained technicians to verify proper operation.