This item is now discontinued. Please browse similar items
Imada SW-1S Data Acquisition Software SKU 5985047
Representative image only.
Imada Mfr # SW-1S – Item # WZ-59850-47
Use buttons on the spreadsheet to operate the gauge from your computer
  • Full SPC and graphing capabilities
  • Calculates statistical data
  • Requires CB-209 cable
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionData Acquisition Software
Two year warranty
Key Features
  • Full SPC and graphing capabilities
  • Calculates statistical data
  • Requires CB-209 cable
  • 2 year warranty
More About this Item
Compatible with Image Force and Torque gauges. Model 59850-47 software and 93955-18 cable are comatoble with models DTX2 or HTG2 gauges. Model 93955-19 cable is compatible with SDS or DSW model gauges.
Use SW-1S data acquisition software to capture and display the current torque values and calculate max/min, average, and standard deviation. Use buttons on the spreadsheet to operate the gauge from your computer. Works with Imada digital torque wrench and screwdriver testers.