This item is now discontinued. Please browse similar items
Imada IHG-C Type C Indentor Height Gauges
Representative image only.
Imada Mfr # IHG-C – Item # WZ-59792-41
Optimize performance of your Imada durometer gauges
  • Compatible with the Imada durometer gauges
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionType C Indentor Height Gauges
  • Warranty2 year
Two year warranty
Key Features
  • Compatible with the Imada durometer gauges
  • 2 year warranty
More About this Item
Choose from a wide range of accessories and replacement parts for your Imada durometer gauges.
Calibration Equipment
Durometers can be tested for proper operation using an indentor height gauge to check the height of the indentor point. Two gauges measure 2, 50 and 100 points. For durometer type Asker C.