This item is now discontinued. Please browse similar items
IKA R078310 Mixing System, Single-Stage In-Line Dispersing Head
Representative image only.
IKA Mfr # U078310 – Item # WZ-04300-01
Allows smooth changeover from laboratory to production plant
  • Small sample requirements for efficient process method determination
  • Eliminates the need to purchase separate mixers—operates as both a batch mixer and in-line mixer
  • Compact, tabletop design minimizes space requirements
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Speed (rpm)3000 to 26,000
  • Power (VAC)220
  • Power (Hz)50
  • DescriptionMixing System, Single-Stage In-Line Dispersing Head
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Small sample requirements for efficient process method determination
  • Eliminates the need to purchase separate mixers—operates as both a batch mixer and in-line mixer
  • Compact, tabletop design minimizes space requirements
  • 1 year warranty
More About this Item
This dispersing and mixing system was developed to meet the needs of research scientists and mixing specialists within the pharmaceutical, cosmetics, chemical, and food industries who need to use the same methods from initial formulation to mass production. The versatile Magic LAB allows you to select from seven different mixing technologies using a variety of available mixing heads, all with programmable speed control to determine the best mixing function for the process. This laboratory machine offers a great variety of applications for many different mixing and particle size reduction processes.
The system consists of a base unit and a single-stage in-line dispersing mixing head; a country-specific plug must be selected separately for operation. By simply pushing buttons on the info center’s display screen, you can easily adjust speed ranges, set nominal and values, and chose your specific language readout (English, Spanish, French, German, or Italian). Depending on the application, the base unit can be modified quickly with optional mixing heads and generators. The modification is simple and does not require any special training or tools (all required tools for standard operation are included).