This item is now discontinued. Please browse similar items
IKA MT 40P.10 Grinding Chamber with Stainless Steel Beater, 40 mL, Bent 45° Angle; 10/pk
Representative image only.
IKA Mfr # 0020017838 – Item # WZ-28711-22
Eliminate contamination with disposable grinding chamber
  • Easily view and clean with transparent, disposable grinding chamber
  • Durable construction suitable for grinding material
SPECS & DESCRIPTION
Specifications & Description
  • Sample Capacity (mL)40
  • MaterialPolypropylene chamber with stainless steel blade
  • DescriptionGrinding Chamber with Stainless Steel Beater, 40 mL, Bent 45° Angle; 10/pk
Key Features
  • Easily view and clean with transparent, disposable grinding chamber
  • Durable construction suitable for grinding material
More About this Item
Choose from a wide range of accessories for the IKA Benchtop Tube Mill Control Mills, items 28710-88 and 28710-89. Mills are sold separately.