IKA 3703000 ULTRA TURRAX Stirring Tube ST-20, (25/pk)
Representative image only.
IKA Mfr # 0003703000 – Item # WZ-04738-06
Choose the sample tube type and size to suit your application
  • Hygienic and clean homogenizing in closed tube system
  • High level of user safety
  • Process sample volumes from 2 - 50 mL
PRODUCT OPTIONS
£377.98GBP / Pkg of 25
£453.58GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)15
  • Capacity (Liters)0.015
  • DescriptionULTRA TURRAX Stirring Tube ST-20, (25/pk)
Key Features
  • Hygienic and clean homogenizing in closed tube system
  • High level of user safety
  • Process sample volumes from 2 - 50 mL
More About this Item
These unique disposable hermetically sealed tubes prevent sample contamination. Each sample tube contains a stirring device, blade, or ball for mixing and homogenizing small samples. Choose from three tube styles with different mixing devices to process your particular sample type.
Stirring Tubes contain a stirring device for gentle mixing, stirring and extraction tasks. Process sample volumes from 2 to 15 mL.