This item is now discontinued. Please browse similar items
IKA 3120900 Separating Funnel Attachment for KS3000 Incubating Shakers
Representative image only.
IKA Mfr # 0003120900 – Item # WZ-51715-49
Expand the uses of your shaker with these easy to use attachments.
  • For shaking out, salting out, extracting, eluting, enriching.
  • The 3 clamping rolls (included in delivery) are height-adjustable for adaption to different separating funnel sizes.
  • The separating funnels are secured with O-rings (5 O-rings included).
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithIKA KS3000 Incubating Shaker
  • Height (in)5.7
  • Width (in)13.1
  • Depth (InchShort)16.7
  • DescriptionSeparating Funnel Attachment for KS3000 Incubating Shakers
Two year warrantyConformite Europeenne
Key Features
  • For shaking out, salting out, extracting, eluting, enriching.
  • The 3 clamping rolls (included in delivery) are height-adjustable for adaption to different separating funnel sizes.
  • The separating funnels are secured with O-rings (5 O-rings included).