This item is now discontinued. Please browse similar items
IKA 0020023172 Wing Style Beater, Tungsten Carbide, 400 mL
Representative image only.
IKA Mfr # 0020023172 – Item # WZ-28711-63
Choose from a variety of vessels and other accessories to expand the capabilities of your IKA® grinding mill
  • Compatible with IKA grinder mill 28711-53
  • Suitable for harder materials up to Mohs hardness 9
  • Compatible with IKA MultiDrive MI 400 (28711-56)
SPECS & DESCRIPTION
Specifications & Description
  • Sample Capacity (mL)400
  • MaterialTungsten carbide
  • DescriptionBeater for Grinding Mill, Tungsten Carbide
Two year warranty
Key Features
  • Compatible with IKA grinder mill 28711-53
  • Suitable for harder materials up to Mohs hardness 9
  • Compatible with IKA MultiDrive MI 400 (28711-56)
More About this Item
Choose from a variety of cutters and beaters to expand the capabilities of your IKA MultiDrive grinding mill