This item is now discontinued. Please browse similar items
Idex XP-218X Flangeless Fitting, Short Head, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
Representative image only.
Idex Mfr # XP-218X – Item # WZ-02014-96
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Natural ETFE
  • 10/PK
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
1 Month Warranty
Includes
Blue ETFE ferrules 02019-33.
Key Features
  • Removable and reusable—saving you money!
  • Natural ETFE
  • 10/PK
  • 1 month warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.