This item is now discontinued. Please browse similar items
Idex VHP-501 Inline VHP Replacement Frit, SS, 0.5 µm; 1/EA
Representative image only.
Idex Mfr # VHP-501 – Item # WZ-02010-13
In-line Solvent Filters for running cleaner systems
  • Help prevent particulates from clogging equipment
  • Parts made of stainless steel and PEEK®
  • Replacement frits available
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialStainless Steel
  • Pore Size (µm)0.5
  • Max Pressure (PSI)15
  • DescriptionInline VHP Replacement Frit, SS, 0.5 µm; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Help prevent particulates from clogging equipment
  • Parts made of stainless steel and PEEK®
  • Replacement frits available
  • 1 year warranty
More About this Item
In-line solvent filters are specifically engineered to help prevent particulate contamination from clogging sensitive equipment. Ideally, they should be placed along the flow path line between the pump and injection valve/autosampler. Available in a variety of porosities, filters threaded to fit 1/16" OD tubing, allowing the use of most standard chromatography high-pressure fittings.