This item is now discontinued. Please browse similar items
Idex V-100L Manual Switching Valve, Right Angle, PEEK/PTFE, 4-Position, 4-Port; 1/EA
Representative image only.
Idex Mfr # V-100L – Item # WZ-02014-87
Easily switch between two fluid paths without disconnecting the lines
  • Enable connections among multiple fluid systems
  • Right angle
  • Swept volume of 6.2 µL
  • 1/4-28 flat-bottom ports for 1/16" OD tubing
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4-28 flat bottom
  • Inlet Port1/4-28 flat bottom
  • Max Pressure (PSI)500
  • Max Pressure (bar)34
  • Wetted MaterialsPEEK/PTFE
  • Tubing OD (in)0.0625
  • Flow PatternRight angle, four-position
  • DescriptionManual Switching Valve, Right Angle, PEEK/PTFE, 4-Position, 4-Port; 1/EA
1 Month Warranty
Includes
Flangeless Black PEEK male nuts (4), flangeless natural ETFE ferrules 02019-74 (4), and black port plug 02013-77 (1).
Key Features
  • Enable connections among multiple fluid systems
  • Right angle
  • Swept volume of 6.2 µL
  • 1/4-28 flat-bottom ports for 1/16" OD tubing
  • 1 month warranty
More About this Item
Manual valves serve well in applications that require operator control or involve lesser frequency of use or injection of smaller sample volumes. Shut off flow during cleaning, column replacement, or other operations without disconnecting the system.