This item is now discontinued. Please browse similar items
Idex UH-904 VHP One-Piece Fitting, Headless, Natural PK, 1/32" OD Tubing, M4 Coned; 1/EA
Representative image only.
Idex Mfr # UH-904 – Item # WZ-02022-62
Fittings for very high pressure applications
  • PK material enables the use of capillary tubing in UHPLC Quick component replacement with minimal down time
  • M4 coned configuration
  • Headless style
  • Requires 4.0 in-lb (0.45 N-m) of torque
  • For 1/32" OD tubing
  • 1/EA
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)M4 coned
  • Compression Size / Tube Od (Port 1) (in)1/32
  • Compression Size / Tube Od (Port 1) (mm)0.794
  • ColorNatural
  • Max Temperature (° F)392
  • Max Temperature (° C)200
  • Max Operating Pressure (PSI)15000
  • DescriptionVHP One-Piece Fitting, Headless, Natural PK, 1/32\" OD Tubing, M4 Coned; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • PK material enables the use of capillary tubing in UHPLC Quick component replacement with minimal down time
  • M4 coned configuration
  • Headless style
  • Requires 4.0 in-lb (0.45 N-m) of torque
  • For 1/32" OD tubing
  • 1/EA
  • 1 year warranty
More About this Item
The most demanding HPLC applications require fittings that will meet higher pressure requirements than standard chromatography fittings can accommodate. They are perfect for use in high-performance analytical systems. VHP PK Fittings are manufactured from a proprietary PEEK blend (PK), which allows use at higher pressures and temperatures.