This item is now discontinued. Please browse similar items
Idex P-411 Port Plug, Microtight® Headless, Black PEEK, 6-32 Coned; 1/EA
Representative image only.
Idex Mfr # P-411 – Item # WZ-02018-38
  • For 6-32 coned port
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPEEK/PCTFE
  • ColorBlack
  • Compatible With6-32 coned port
  • DescriptionPort Plug, Microtight® Headless, Black PEEK, 6-32 Coned; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • For 6-32 coned port
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications.