This item is now discontinued. View the direct replacement: WZ-12031-20 or browse similar items
Idex P-316 Port Plug, Standard Knurl, Natural PFA, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Mfr # P-316 – Item # WZ-02013-79
Available in various thread sizes and materials
  • Easily identify columns with color coded plugs
  • For 1/4-28 flat-bottom port
  • Natural PFA construction
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPFA
  • ColorNatural
  • Compatible With1/4-28 flat-bottom port
  • DescriptionP-316 Port Plug, Standard Knurl, Natural PFA, 1/4-28 Flat-Bottom; 1/EA
1 Month Warranty
Key Features
  • Easily identify columns with color coded plugs
  • For 1/4-28 flat-bottom port
  • Natural PFA construction
  • 1 month warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.