Idex Super Flangeless™ Nut, Standard Knurl, Red PEEK, 1/32" or 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Mfr # P-255R – Item # WZ-02021-66
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Standard knurl
  • Red PEEK
  • 1 year warranty
PRODUCT OPTIONS
£6.77GBP / Each
£8.12GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (in)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/32
  • ColorRed
  • DescriptionSuper Flangeless™ Nut, Standard Knurl, Red PEEK, 1/32\" or 1/16\" OD Tubing, 1/4-28 Flat Bottom; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Standard knurl
  • Red PEEK
  • 1 year warranty
More About this Item
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
Pressure rating of nut depends on ferrule used.