This item is now discontinued. Please browse similar items
Idex OC-416 Omega Frit with Ring, PEEK, 2 µm, Green; 1/EA
Representative image only.
Idex Mfr # OC-416 – Item # WZ-42718-76
Extend filtration life and consistency with replaceable frits
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
  • Strong and resilient
  • Sealing ring made of PEEK
  • Enhanced sealing properties and stronger resistance to chemicals
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPEEK
  • Pore Size (µm)2
  • DescriptionOmega Frit with Ring, PEEK, 2 µm, Green
  • Warranty1 year
1 Year Warranty
Key Features
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
  • Strong and resilient
  • Sealing ring made of PEEK
  • Enhanced sealing properties and stronger resistance to chemicals
  • 1 year warranty
More About this Item
Easily replaceable frits save you time and money lost to frit-cleaning labor, plus eliminate the problem of embedded debris re-entering your instrument. These frits are available in the most common HPLC filtration ratings. Each includes a PCTFE or PEEK polymer sealing ring, which are slightly thicker than the frit disc for extra sealing and chemical resistance. Many also have the most common outer diameters — 0.250" and 0.254" — making them ideal for use in inline or precolumn filters.
These frits offer stellar uniform porosity, which in turn, ensures a longer filtration life.