Idex Low Pressure Chromatography Unions, Tees, and Crosses Fitting Kit
Representative image only.
Idex Mfr # 1457 – Item # WZ-02008-50
Save time and money with a fitting kits
  • Constructed of chemically inert PEEK plastic
  • Limited Lifetime Warranty
£608.13GBP / Each
£729.76GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Descriptionlow-pressure unions, tees, crosses and kits; fitting kit
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Constructed of chemically inert PEEK plastic
  • Limited Lifetime Warranty
More About this Item
PEEK Fitting Kit contains twenty ferrules, six FINGERTIGHT III nuts with ferrules, two metric adapters, two 10-32 female to 1/4-28 female adapters, two unions with 0.020" bore, one union with 0.050" bore, tees with 0.020" bore and 0.050" bore each, and crosses with 0.020" and 0.050" bore each.