This item is now discontinued. Please browse similar items
Idex LiteTouch and Super Flangeless Ferrule Removal Tool; 1/EA
Representative image only.
Idex Mfr # LT-300 – Item # WZ-01939-39
Secure a leak-tight connection
  • Wrenches for tightening chromatography fittings
  • Specifically designed to remove LifeTouch® and Super Flangeless™ ferrules from tubing
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSteel/plastic handle
  • Compatible WithLifeTouch and Super Flangeless ferrules
  • DescriptionLiteTouch and Super Flangeless Ferrule Removal Tool; 1/EA
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Wrenches for tightening chromatography fittings
  • Specifically designed to remove LifeTouch® and Super Flangeless™ ferrules from tubing
  • Limited Lifetime Warranty
More About this Item
Durable and reliable wrench set comes in handy for your chromatography needs.
Simply slide the blade slot between the lock ring and the ferrule body. With a slight twist, the ring will pop off, releasing the ferrule from the tubing.