This item is now discontinued. Please browse similar items
Idex Fingertight Two-Piece, Double-Wing, Natural PEEK, 1/16" OD Tubing, 1/4-28 Coned; 1/EA
Representative image only.
Idex Mfr # F-301 – Item # WZ-02007-02
Withstands up to 6000 psi!
  • Winged nut head provides extra leverage when tightening into a receiving port
  • Double wing
  • 1/4-28 coned
  • 1/EA
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • Max Operating Pressure (PSI)6000
  • DescriptionFingertight Two-Piece, Double-Wing, Natural PEEK, 1/16\" OD Tubing, 1/4-28 Coned; 1/EA
Key Features
  • Winged nut head provides extra leverage when tightening into a receiving port
  • Double wing
  • 1/4-28 coned
  • 1/EA
More About this Item
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.