This item is now discontinued. Please browse similar items
Idex F-160X Fingertight Two-Piece Fitting, Standard Knurl, SS, 1/16" OD Tubing, 1/4-28 Coned, 10/PK
Representative image only.
Idex Mfr # F-160X – Item # WZ-01939-12
Popular threaded coned port configurations support quality high-pressure connections
  • Perfect for analytical instruments, in-vitro diagnostics, electronics and other biotechnology applications
  • 1/4-28 coned
  • 10/PK
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialSS/PEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (in)1/4-28 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorGray
  • Max Operating Pressure (PSI)5000
  • DescriptionF-160X Fingertight Two-Piece, Standard Knurl, SS, 1/16\" OD Tubing, 1/4-28 Coned, 10/PK
Key Features
  • Perfect for analytical instruments, in-vitro diagnostics, electronics and other biotechnology applications
  • 1/4-28 coned
  • 10/PK
More About this Item
Two-piece fittings comprise of a separate nut and ferrule. Save money by replacing only the ferrule. For mass spectrometer electrospray application order ferrule 02019-15.