Idex Inlet Solvent Filter, SS/PCTFE, 10 µm, 1/8" OD Tubing; 1/EA
Representative image only.
Idex Mfr # A-240 – Item # WZ-42711-14
Keep damaging particles out of your system with inlet solvent filters
  • Protect your system from particulate matter that can cause damage
  • Filters available to fit specific system specifications
  • Easy to replace without tools
  • Rubber tipped temples offer secure fit
  • Soft, rubber nosepiece for all-day comfort
  • 1 year warranty
PRODUCT OPTIONS
£59.30GBP / Each
£71.16GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialStainless Steel
  • Housing MaterialPCTFE
  • Pore Size (µm)10
  • Connections1/4-28 coned
  • DescriptionInlet Solvent Filter, SS/PCTFE, 10 µm, 1/8\" OD Tubing; 1/EA
  • Warranty1 year
1 Year Warranty
Includes
One P-100 fingertight fitting
Key Features
  • Protect your system from particulate matter that can cause damage
  • Filters available to fit specific system specifications
  • Easy to replace without tools
  • Rubber tipped temples offer secure fit
  • Soft, rubber nosepiece for all-day comfort
  • 1 year warranty
More About this Item
For good preventive maintenance, inlet solvent filters are an optimal way to cleanse solvents, avoiding pump cavitation and system damage. Different style stainless steel filters are available for specific system specifications and can be easily replaced regularly without tools.
To be used with 1/8" OD tubing.