Idex Rotor Seal for Valve Model 9010, ETFE; 1/EA
Representative image only.
Idex Mfr # 9010-051 – Item # WZ-02015-74
Replace rotor seals for longer valve life
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve model 9010
  • ETFE seal
  • 1 year warranty
PRODUCT OPTIONS
£123.35GBP / Each
£148.02GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialETFE
  • Compatible WithValve model 9010
  • DescriptionRotor Seal for Valve Model 9010, ETFE; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve model 9010
  • ETFE seal
  • 1 year warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.