This item is now discontinued. Please browse similar items
Idex 79801-999 RheBuild® Kit for Actuated Valve 02013-61; 1/EA
Representative image only.
Idex Mfr # 79801-999 – Item # WZ-02013-19
Regular maintenance ensures optimal valve performance
  • Optimize performance of your rotary shear valves
  • Use with valve 02013-61
  • 1 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithActuated valve 02013-61
  • DescriptionRheBuild® Kit for Actuated Valve 02013-61; 1/EA
1 Month Warranty
Key Features
  • Optimize performance of your rotary shear valves
  • Use with valve 02013-61
  • 1 month warranty
More About this Item
Routine valve maintenance using RheBuild® kits or rapid replacement pods ensure optimal performance. The entire pod can be replaced quickly as a unit with virtually no instrument downtime for repair.