This item is now discontinued. Please browse similar items
Idex Sample Loop for VHP, SS, 5 µL, 1/16" OD; 1/EA
Representative image only.
Idex Mfr # 7755-200 – Item # WZ-02016-61
Specifically designed for VHP applications
  • Square-cut, burr-free ends provide a firm, flush connection to valve ports
  • Constructed from 316 stainless steel
  • High pressure resistance, low permeability
  • 5 µL sample volume
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Wetted Materials316 stainless steel
  • Tubing Outside Diameter (OD) (in)0.0625
  • Total Volume Capacity (µL)5
  • DescriptionSample Loop for VHP, SS, 5 µL, 1/16\" OD; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Square-cut, burr-free ends provide a firm, flush connection to valve ports
  • Constructed from 316 stainless steel
  • High pressure resistance, low permeability
  • 5 µL sample volume
  • 1 year warranty
More About this Item
A sample loop is a defined length of tubing and fittings configured to match the angle of the injection valve ports. A sample is loaded and held in the loop until injection is triggered, either automatically or manually. Stainless steel loops are supplied with unswaged fittings, so the tube can be completely bottomed in the injection port before the ferrule is swaged. The loop volumes are nominal.