This item is now discontinued. Please browse similar items
Idex Rotor Seal for Valve Models 7060/7066, Vespel®; 1/EA
Representative image only.
Idex Mfr # 7060-070 – Item # WZ-02015-57
Replace rotor seals for longer valve life
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 7060/7066
  • Vespel® seal
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialVespel®
  • Compatible WithValve models 7060/7066
  • DescriptionRotor Seal for Valve Models 7060/7066, Vespel®; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 7060/7066
  • Vespel® seal
  • 1 year warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.