This item is now discontinued. Please browse similar items
Idex 6000-278 RheFlex® Two-Piece Finger-Tight Fitting, 1/8" OD Tubing, Flangeless; 10/PK
Representative image only.
Idex Mfr # 6000-278 – Item # WZ-02032-28
Simplify by getting an all-in-one fitting for your fluidic needs
  • PEEK construction for an inert, biocompatible connection
  • Color-coded ChromTRAC knobs for easy identification
  • Fits 1/8" OD Tubing Comes in a 10 Pack
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPEEK
  • Compression Size / Tube Od (Port 1) (in)1/8
  • DescriptionRheFlex® Two-Piece Finger-Tight Fitting, 1/8\" OD Tubing, Flangeless; 10/PK
  • Warranty1 year
1 Year Warranty
Key Features
  • PEEK construction for an inert, biocompatible connection
  • Color-coded ChromTRAC knobs for easy identification
  • Fits 1/8" OD Tubing Comes in a 10 Pack
  • 1 year warranty
More About this Item
A. One-Piece RheFlex M4 Fitting
Eliminates the need for separate nuts and ferrules
The combination of a one-piece nut and ferrule makes connections easier and faster. The fitting features a unique seal-tight grip. When used with PEEK tubing sleeve, the fitting can hold up to 5000 psi (345 bar). They are compatible with ChromTRAC knobs.
B. Two-Piece RheFlex Finger-Tight Fittings
No need for tools to tighten these fittings
Included ChromTRAC knobs allow for quick identification. The 10-32 coned bottom fittings are available in standard, short and extra-long lengths. Each fitting comes with a PEEK ferrule. The fittings are rated for use up to 7000 psi (483 bar).