Idex Rotor Seal for Valve Models 3725i/3725i-038, PEEK; 1/EA
Representative image only.
Idex Mfr # 3725-018 – Item # WZ-02015-29
Replace rotor seals for longer valve life
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 3725i/3725i-038
  • PEEK seal
  • 1 year warranty
PRODUCT OPTIONS
£174.59GBP / Each
£209.51GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPEEK
  • Compatible WithValve models 3725i/3725i-038
  • DescriptionRotor Seal for Valve Models 3725i/3725i-038, PEEK; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 3725i/3725i-038
  • PEEK seal
  • 1 year warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.