This item is now discontinued. Please browse similar items
Idex 1593-01 VHP Prep Union Body, ZDV, SS, 0.050" ID, 1/8" OD Tubing, 1/4-28 Coned; 1/EA
Representative image only.
Idex Mfr # 1593-01 – Item # WZ-02024-16
Zero dead volume unions ensure accurate analysis
  • Precision machined from 316 stainless steel to withstand high pressures
  • Securely join fluid pathway components
  • Holds tight connection even through vibration
  • VHP ZDV union body
  • 1/4-28 coned configuration
  • 0.050" thru-hole diameter
  • Swept volume of 1.48 µL
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialStainless Steel
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 1) (in)1/4-28 Coned
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorNatural
  • DescriptionVHP Prep Union Body, ZDV, SS, 0.050\" ID, 1/8\" OD Tubing, 1/4-28 Coned; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Precision machined from 316 stainless steel to withstand high pressures
  • Securely join fluid pathway components
  • Holds tight connection even through vibration
  • VHP ZDV union body
  • 1/4-28 coned configuration
  • 0.050" thru-hole diameter
  • Swept volume of 1.48 µL
  • 1 year warranty
More About this Item
These Idex® VHP stainless steel ZDV unions provide a tight, secure connection in your chromatography fluid path.