Idec PS5R-B24 DIN-Rail Power Supply 15W (24 VDC, 0.6 A)

Representative image only.
Idec Mfr # PS5R-VB24 – Item # WZ-26901-06
Compact design provides exceptional power output for all system needs
  • Save valuable DIN-rail space while filling your power requirements

PRODUCT OPTIONS

£79.00GBP / Each
£94.80GBP (incl VAT)

£107.00GBP / Each
£128.40GBP (incl VAT)

for an additional
£107.00GBP / Each
£128.40GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Output Voltage24 VDC
  • Output Current0.6 A
  • Ripple and Noise≤ 2%
  • Line Regulation - Current≤ 0.4%
  • Line Regulation - Voltage≤ 0.4%
  • Load Regulation - Voltage1.5% max
  • Width (in)1.75
  • Depth (in)3.75
  • Height (in)3
  • Power Rating (watts)15
  • Power (VAC)100 to 240
  • DescriptionDIN-Rail Power Supply 15W (24 VDC, 0.6 A)

Key Features

  • Save valuable DIN-rail space while filling your power requirements

More About this Item

These switching power supplies offer the versatility of DIN-rail or direct surface mounting. The unique spring-up, terminals feature an IP20 (finger-safe) rating — ideal for ring lug terminated wires. Adjust output voltage with easy access trim pot to +/-10%. Outputs also feature auto-resetting overcurrent protection and 2% regulated maximum ripple. Other features include UL508 listing (no derating), fused inputs and an LED verifying power.
Models 26901-08, -10, -12, and –16 provide UL1604 listing for hazardous locations and Class 2 rating (-08, -10, and –12). Models 26901-14, -18, and –20 offer the capability of being connected in parallel to increase the current output.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At
Live chat