This item is now discontinued. View the direct replacement: WZ-99540-57 or browse similar items
I-Chem 341-1250 pre-cleaned sample jars with certificate of analysis, 1250 mL, 70 mm cap
Representative image only.
I-Chem Mfr # 341-1250 – Item # WZ-34701-40
Often bought with…
Cole-Parmer® Essentials Glass Erlenmeyer Flasks, Graduated, Heavy Rim
£13.20 - £49.47GBP / Pkg of 8
£15.84 - £59.36GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (L)1.25
  • Capacity (oz)40
  • MaterialGlass
  • Cap MaterialPTFE-lined Polypropylene
  • Cap / Stopper Size70 mm
  • Bottle ColorAmber
  • Descriptionpre-cleaned sample jars with certificate of analysis, 1250 mL, 70 mm cap
More About this Item
Protect UV-light sensitive soil, sludge, and sediment samples in these sample jars. Containers are composed of type III glass and include a PTFE-lined, polypropylene cap. Jars are pre-cleaned to meet Environmental Protection Agency (EPA) Performance Based Specifications for metals and semi-volatiles, pesticides, and PCBs analysis. I-CHEM is an ISO9001 Registered Supplier.