Hydrion 7200 Plastic pH Strips, Range 0.0 to 6.0
Representative image only.
Hydrion Mfr # 7200 – Item # WZ-59200-12
Three times the matching area compared with other plastic strips
  • Easier to read and color match than other plastic strips
£20.08GBP / Each
£24.10GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • TestpH
  • Measurement Points0 to 6 pH in 0.5 pH unit increments
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionPlastic pH Strips, Range 0.0 to 6.0
Key Features
  • Easier to read and color match than other plastic strips
More About this Item
Immediate pH testing is available in brilliant, long lasting colors. These direct-reading pH papers give a simple, single color match for all pH values in distinct, bright color separations. Clear plastic cases contain 100 dip sticks and feature a handy matching color chart.