HTL 2254 Clinipet Series CP500 Fixed-Volume Pipettor, Blue; 500 µL
Representative image only.
Mfr # 2254 – Item # WZ-24551-28
Enjoy accuracy and precision with durable, UV-resistant and autoclavable pipettors
  • Easily removable tip ejector makes it simple to work with narrow tubes
  • Low pipetting forces prevent repetitive strain injury
  • Unique tip sealing system
  • Available in 33 models from 2 µL to 1000 µL
  • 3 year warranty
PRODUCT OPTIONS
£73.78GBP / Each
£88.54GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • AutoclavableYes
  • Max Volume (µL)500
  • Accuracy (%)±0.6
  • Precision (%)±0.2
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeBlue
  • DescriptionFixed-Volume Pipettor, Blue; 500 µL
  • Warranty3 year
3 Year Warranty
Key Features
  • Easily removable tip ejector makes it simple to work with narrow tubes
  • Low pipetting forces prevent repetitive strain injury
  • Unique tip sealing system
  • Available in 33 models from 2 µL to 1000 µL
  • 3 year warranty
More About this Item
Whether you're right- or left-handed, the Clinipet+ fixed-volume pipettor is smooth to use, thanks to a sliding tip ejector button and smartly designed handle. Durable PVDF shaft guarantees optimum mechanical and chemical resistance. These fixed-volume pipettors also ensure accurate pipette calibration.