This item is now discontinued. Please browse similar items
Honeywell Rig Dog™ Xtreme Impact ANSI A6 Cut-Resistant Glove, Slipon Cuff, 9/L
Representative image only.
Honeywell Mfr # 42-612BY/9L – Item # WZ-86326-54
Enhanced grip when handling dry and oily surfaces
  • ANSI A6 (ISO level F) cut resistance; level 4 abrasion resistance Excellent impact protection Rapid sweat absorption promotes comfort on long shifts Slip-on cuff for easy donning
  • Size large
SPECS & DESCRIPTION
Specifications & Description
  • Size9/L
  • Length (in)10
  • Length (cm)25.4
  • ColorRed with green trim
  • ApprovalANSI A6 (ISO level F) cut resistance, level 4 abrasion resistance
  • Max Temperature (° F)86
  • Max Temperature (° C)30
  • DescriptionImpact ANSI A6 Cut-Resistant Glove, Slipon Cuff, 9/L
Key Features
  • ANSI A6 (ISO level F) cut resistance; level 4 abrasion resistance Excellent impact protection Rapid sweat absorption promotes comfort on long shifts Slip-on cuff for easy donning
  • Size large
More About this Item
Rig Dog™ Xtreme gloves from Honeywell put safety first. Protecting workers across tough environments, they are well-fit for rigging, drilling, material handling, ship building and other rugged industrial applications. The cut-resistant and enhanced oil-grip palm--featuring minimum stitching--enables these gloves to be worn with comfort and confidence all day long.
These gloves are manufactured from reinforced thermoplastic rubber, part of the CoreNest™ technology, woven into each pair for safety and efficiency. Enhanced back-of-hand impact protection helps prevent harm to knuckles, finger tips and thumb crotch.