This item is now discontinued. Please browse similar items
Honeywell Servus Polyurethane Processing Overboots, XS; 1 Pair
Representative image only.
Honeywell Mfr # IT12PT17-BLU-XSM – Item # WZ-86326-71
Protect yourself against abrasions, slipping, and splashes
  • Completely waterproof Chemical-resistant polyurethane construction Light weight prevents foot and leg fatigue Easily put on and take off Unique strap system provides a secure fit to stop boot from slipping Available in mens’ sizes XS to 3XL
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)12
  • ColorWhite & blue
  • Men's SizeXS
  • Thickness (mil)8
  • DescriptionPolyurethane Processing Overboots, XS
Key Features
  • Completely waterproof Chemical-resistant polyurethane construction Light weight prevents foot and leg fatigue Easily put on and take off Unique strap system provides a secure fit to stop boot from slipping Available in mens’ sizes XS to 3XL
More About this Item
Servus® by Honeywell’s™ processing overboots are constructed of heavy-duty 0.8 mm thermal polyurethane to keep your feet dry. The 100% waterproof design offers splash protection against water and most chemicals for longer wear. Easily put on and take off using the extra-large gusset. The one-of-a-kind open tread design allows for firm footing in wet and oily conditions. The ankle strap provides a snug, adjustable fit for your needs, and its snap closure allows for easy sanitation. Blue and white color; with soft toe.