This item is now discontinued. Please browse similar items
Honeywell 31-30135 Laser Eyewear, Helium Neon/Red Diodes
Representative image only.
Honeywell Mfr # 31-30135 – Item # WZ-86438-29
Excellent visible light transmission minimizes distortion
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBlack
  • Lens ColorBlue
  • Laser TypeHelium Neon (HeNe), red diodes
  • ComplianceANSI Z136.1-1986, Z87.1-2003
  • WavelengthOD 1-2 at 630-680 nm
  • DescriptionLaser Eyewear, Helium Neon/Red Diodes
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
These lightweight, impact-resistant polymer lenses are suitable for low-powered lasers or indirect laser exposure. Wavelength, optical density, and lot number are inscribed across lens for easy identification. All eyewear comes with protective case, cleaning cloth, and safety hanging cord.
LGF Series
LGF eyewear features polymer filter that protect against femtosecond (fs) and picosecond (ps) lasers. These polymer filters maintain their optical density for 10 seconds according to independent third-party testing and international standards. Eyewear features one-piece, impact-resistant polymer lens with integral molded side shields and offers a large protective area with unrestricted wide-angle viewing. Contoured frame conforms to the forehead providing excellent orbital coverage. Fits over most prescription frames.
Compliance: ANSI Z136.1-1986, Z87.1-2003 (glasses unsuitable for direct laser radiation)