Honeywell 04973-XX1-333-20-00-000 conductivity probe, K=1.0
Representative image only.
Honeywell Mfr # 04973-XX1-333-20-00-000 – Item # WZ-56611-32
Optimize performance of your Honeywell conductivity control system
  • Keep your conductivity control system running properly with these accessories
  • 1 year warranty
PRODUCT OPTIONS
£601.37GBP / Each
£721.64GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolysulfone, Graphite
  • Compatible WithHoneywell controller
  • Connections3/4\" NPT(M)
  • Length (in)240
  • Descriptionconductivity probe, K=1.0
  • Warranty1 year
1 Year Warranty
Key Features
  • Keep your conductivity control system running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Honeywell conductivity control system.
Conductivity electrode, 0 to 199.9 uS range, K = 1.0. Polysulfone and graphite electrode with 20-ft (6.1-m) cable. 3/4" NPT(M) process connections.