This item is now discontinued. Please browse similar items

Hirschmann opus Titrator, 10 mL; 100 to 240 VAC, 50/60 Hz

Representative image only.
Hirschmann Mfr # 9582010 – Item # WZ-25503-04
Obtain precise titration results with electronic control and dispensing
  • Configure titration parameters on user-friendly TFT touch screen
  • Time-saving rapid titration in conjunction with fine titration ensures system accuracy
  • Save up to nine procedures and settings with on-board memory for quick recall
  • Easily transfer data via RS-232 and USB interfaces
  • Titration range from 0.01 to 10.0 mL
  • Velocity rate in 16 stages from 0.01 mL/s to 2 mL/s
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Display Type3.5\" TFT Touchscreen
  • Capacity (mL)10
  • Dispensing AccuracyR=0.2%; CV=0.07%
  • OutputRS-232 and USB
  • Data LoggingUp to 9 configurations
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Length (in)5 1/10
  • Width (in)4 19/64
  • Height (in)2 1/5
  • Length (cm)13
  • Width (cm)11
  • Height (cm)5.5
  • DescriptionTitrator, 10 mL; 100 to 240 VAC, 50/60 Hz
  • Warranty1 year
1 Year Warranty

Key Features

  • Configure titration parameters on user-friendly TFT touch screen
  • Time-saving rapid titration in conjunction with fine titration ensures system accuracy
  • Save up to nine procedures and settings with on-board memory for quick recall
  • Easily transfer data via RS-232 and USB interfaces
  • Titration range from 0.01 to 10.0 mL
  • Velocity rate in 16 stages from 0.01 mL/s to 2 mL/s
  • 1 year warranty

More About this Item

The electronic titration system is user-friendly with an intuitive touch screen and direct titration from the laboratory bottle. Titration impulse triggered by the motor allows for droplet titration of 1 droplet per button push. Prevent reagent loss with simple and rapid venting, made possible with the media recirculating system.
Advanced safety and protection features are valuable for daily routines. Protect your data and procedures from unauthorized personnel using the PIN function. The PIN function can also enable only select functions to be performed by selected users. Freely rotate the titrator on the reagent bottle, to ensure the bottle label is always pointing towards the user. Your light-sensitive samples will be protected by the light protection window, included with your titration system.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat